Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515599_m1
          See other CTSW GE Assays ›
          Gene Symbol:
          Ctsw
          Gene Name:
          cathepsin W
          Gene Aliases:
          lymphopain
          Chromosome Location:
          Chr.19: 5465240 - 5470387 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515599_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsw
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009985.4 NP_034115.2 2 - 3 179 84
          GenBank mRNA AF014941.1 - 2 - 3 164 84
          AK088375.1 - 2 - 3 180 84
          AK163626.1 - 2 - 3 183 84
          BB850740.1 - 2 - 3 180 84
          BC019136.1 - 2 - 3 180 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000967.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsw

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          immune response
          proteolysis involved in cellular protein catabolic process
          cysteine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928903 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021981 AK088375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021981 AK163626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021981 BC019136.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021981 BE447240.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021981 NM_009985.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552939 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162404 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021981 AK088375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021981 AK163626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021981 BC019136.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021981 BE447240.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021981 NM_009985.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P264924 AK088375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P264924 AK163626.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P264924 BC019136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P264924 BE447240.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P264924 NM_009985.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162404 - -
          Whole Mouse Genome G4122A A_52_P264924 AK088375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P264924 AK163626.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P264924 BC019136.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P264924 BE447240.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P264924 NM_009985.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120551 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103526 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162404 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14261503 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14261506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14261502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14304215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14261507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14403970 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14504851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14403967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14403970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928871 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465070 - -
          Affymetrix HT MG-430 PM Array Plate 1422632_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465059 - -
          Mouse Genome 430 2.0 Array 1422632_at - -
          Mouse Genome 430 2.0 Array 1422632_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13041_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13041_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900008613.mm.1 NM_009985.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008613.mm.1 TC1900000967.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008612.mm.1 NM_009985.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008612.mm.1 TC1900000967.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008611.mm.1 NM_009985.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008611.mm.1 TC1900000967.mm.1 MTA TC
          Mouse Expression Set 430 1422632_PM_at - -
          Mouse Expression Set 430 1422632_at - -
          Murine Genome U74v2 Set 92214_PM_at - -
          Murine Genome U74v2 Set 92214_at - -
          Mouse Genome 430A 2.0 Array 1422632_at - -
          Mouse Genome 430A 2.0 Array 1422632_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422632_at - -
          Affymetrix HT Mouse Genome 430A Array 1422632_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 447264 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 360368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721399 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721399 - -
          mouse-6 v1.1 (Illumina) scl53496.8.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 360368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53496.8.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2721399 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline