Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00515900_m1
          See other GSTZ1 GE Assays ›
          Gene Symbol:
          Gstz1
          Gene Name:
          glutathione transferase zeta 1 (maleylacetoacetate isomerase)
          Gene Aliases:
          MAAI
          Chromosome Location:
          Chr.12: 87147165 - 87164723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00515900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gstz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252555.1 NP_001239484.1 9 - 10 952 76
          NM_001252556.1 NP_001239485.1 7 - 8 581 76
          NM_010363.4 NP_034493.1 8 - 9 649 76
          GenBank mRNA AB041613.1 - 9 - 10 952 76
          AF093418.1 - 8 - 9 598 76
          AK002398.1 - 8 - 9 606 76
          AK012796.1 - 8 - 9 650 76
          AK075927.1 - 8 - 9 603 76
          BC031777.1 - 8 - 9 638 4168439 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000869.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gstz1

          Panther Classification:

          Molecular Function -

          isomerase

          Gene Ontology Categories:

          Function(s) Process(es)

          L-phenylalanine catabolic process
          tyrosine catabolic process
          glutathione metabolic process
          metabolic process
          aromatic amino acid family metabolic process
          catalytic activity
          glutathione transferase activity
          maleylacetoacetate isomerase activity
          transferase activity
          isomerase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614427 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988708 AK075927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988708 BC031777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988708 AF093418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988708 NM_010363.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31965 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174073 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105921 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988708 AK075927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988708 BC031777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988708 AF093418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988708 NM_010363.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P499879 AK075927.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P499879 BC031777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P499879 AB041613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P499879 AF093418.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P499879 AK002398.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28614421 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174073 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02570997 - -
          Whole Mouse Genome G4122A A_52_P499879 AK075927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499879 BC031777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499879 AB041613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499879 AF093418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499879 AK002398.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120870 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105921 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122727 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122722 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10122741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614427 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28614429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28614383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09961078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10003871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10122723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09957758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10063547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09936947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10052865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10063550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28614470 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10397514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397517 - -
          Affymetrix HT MG-430 PM Array Plate 1427552_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10397507 - -
          Mouse Genome 430 2.0 Array 1427552_a_at - -
          Mouse Genome 430 2.0 Array 1427552_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14874_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14874_at - -
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_001252555.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_001252555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_001252556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_001252556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_010363.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 NM_010363.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006374.mm.1 TC1200000869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_001252555.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_001252555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_001252556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_001252556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_010363.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 NM_010363.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006373.mm.1 TC1200000869.mm.1 MTA TC
          Mouse Expression Set 430 1427552_PM_a_at - -
          Mouse Expression Set 430 1427552_a_at - -
          Murine Genome U74v2 Set 160350_PM_at - -
          Murine Genome U74v2 Set 160350_at - -
          Mouse Genome 430A 2.0 Array 1427552_a_at - -
          Mouse Genome 430A 2.0 Array 1427552_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427552_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427552_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000451 - -
          Illumina mouse-6 v1.0 expression beadchip 7000451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229964 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229964 - -
          mouse-6 v1.1 (Illumina) scl42938.12_138-S - -
          Illumina Sentrix mouse-6.v1 bead array 7000451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42938.12_138-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229964 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229964 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl42938.12_138-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline