Search
Search

Hdac3| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010411.2 | NP_034541.2 | 1 - 2 | 137 | 58 | |
| GenBank mRNA | AF074881.1 | - | 1 - 2 | 84 | 58 | |
| AF074882.1 | - | 1 - 2 | 84 | 58 | ||
| AF098295.1 | - | 1 - 2 | 62 | 58 | ||
| AF125536.1 | - | 1 - 2 | 69 | 58 | ||
| AK145158.1 | - | 1 - 2 | 103 | 58 | ||
| BC139300.1 | - | 1 - 2 | 165 | 8862322 | 58 | |
| BC139301.1 | - | 1 - 2 | 165 | 8862323 | 58 | |
| BQ932476.1 | - | 1 - 2 | 149 | 58 | ||
| CJ166955.1 | - | 1 - 2 | 140 | 58 | ||
| JN651901.1 | - | 1 - 2 | 67 | 58 | ||
| JN651902.1 | - | 1 - 2 | 67 | 58 | ||
| JN651903.1 | - | 1 - 2 | 67 | 58 | ||
| JN651904.1 | - | 1 - 2 | 67 | 58 | ||
Set Membership: |
|