Search
Search

Hdac6| Gene Symbol: Hdac6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130416.1 | NP_001123888.1 | 19 - 20 | 2057 | 59 | |
| NM_010413.3 | NP_034543.3 | 19 - 20 | 2132 | 59 | ||
| GenBank mRNA | AF006603.2 | - | 18 - 19 | 1817 | 59 | |
| AK148092.1 | - | 19 - 20 | 1978 | 59 | ||
| AK148144.1 | - | 19 - 20 | 1948 | 59 | ||
| AK154099.1 | - | 19 - 20 | 1996 | 59 | ||
| AK155545.1 | - | 17 - 18 | 3030 | 59 | ||
| AK155546.1 | - | 19 - 20 | 1996 | 59 | ||
| BC033428.1 | - | 9 - 10 | 908 | 59 | ||
| BC041105.1 | - | 19 - 20 | 1989 | 5352032 | 59 | |
Set Membership: |
|