Search
Search

Kcnma1| Gene Symbol: Kcnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253358.1 | NP_001240287.1 | 27 - 28 | 3338 | 68 | |
| NM_001253359.1 | NP_001240288.1 | 27 - 28 | 3338 | 68 | ||
| NM_001253360.1 | NP_001240289.1 | 26 - 27 | 3329 | 68 | ||
| NM_001253361.1 | NP_001240290.1 | 28 - 29 | 3443 | 68 | ||
| NM_001253362.1 | NP_001240291.1 | 27 - 28 | 3434 | 68 | ||
| NM_001253363.1 | NP_001240292.1 | 27 - 28 | 3434 | 68 | ||
| NM_001253364.1 | NP_001240293.1 | 26 - 27 | 3422 | 68 | ||
| NM_001253365.1 | NP_001240294.1 | 26 - 27 | 3257 | 68 | ||
| NM_001253366.1 | NP_001240295.1 | 26 - 27 | 3257 | 68 | ||
| NM_001253367.1 | NP_001240296.1 | 25 - 26 | 3248 | 68 | ||
| NM_001253368.1 | NP_001240297.1 | 27 - 28 | 3347 | 68 | ||
| NM_001253369.1 | NP_001240298.1 | 27 - 28 | 3341 | 68 | ||
| NM_001253370.1 | NP_001240299.1 | 26 - 27 | 3335 | 68 | ||
| NM_001253371.1 | NP_001240300.1 | 27 - 28 | 3338 | 68 | ||
| NM_001253372.1 | NP_001240301.1 | 26 - 27 | 3335 | 68 | ||
| NM_001253373.1 | NP_001240302.1 | 25 - 26 | 3266 | 68 | ||
| NM_001253374.1 | NP_001240303.1 | 25 - 26 | 3095 | 68 | ||
| NM_001253375.1 | NP_001240304.1 | 25 - 26 | 3248 | 68 | ||
| NM_001253376.1 | NP_001240305.1 | 25 - 26 | 3248 | 68 | ||
| NM_001253377.1 | NP_001240306.1 | 25 - 26 | 3248 | 68 | ||
| NM_001253378.1 | NP_001240307.1 | 25 - 26 | 3248 | 68 | ||
| NM_010610.3 | NP_034740.2 | 26 - 27 | 3422 | 68 | ||
| GenBank mRNA | AF156674.1 | - | 26 - 27 | 3147 | 68 | |
| AF465244.1 | - | 25 - 26 | 2973 | 68 | ||
| AK161962.1 | - | 5 - 6 | 616 | 68 | ||
| BC065068.1 | - | 27 - 28 | 3435 | 68 | ||
| BC128331.1 | - | 26 - 27 | 2978 | 68 | ||
| FJ872117.1 | - | 27 - 28 | 3063 | 68 | ||
| HQ221743.1 | - | 27 - 28 | 3063 | 68 | ||
| HQ221744.1 | - | 25 - 26 | 2820 | 68 | ||
| HQ221745.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221746.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221747.1 | - | 26 - 27 | 2982 | 68 | ||
| HQ221748.1 | - | 26 - 27 | 3054 | 68 | ||
| HQ221749.1 | - | 27 - 28 | 3063 | 68 | ||
| HQ221750.1 | - | 26 - 27 | 2982 | 68 | ||
| HQ221751.1 | - | 27 - 28 | 3069 | 68 | ||
| HQ221752.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221753.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221754.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221755.1 | - | 26 - 27 | 3060 | 68 | ||
| HQ221756.1 | - | 27 - 28 | 3159 | 68 | ||
| HQ221757.1 | - | 28 - 29 | 3168 | 68 | ||
| HQ221758.1 | - | 26 - 27 | 3147 | 68 | ||
| HQ221760.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221761.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ221762.1 | - | 26 - 27 | 3060 | 68 | ||
| HQ221763.1 | - | 25 - 26 | 2973 | 68 | ||
| HQ529469.1 | - | 26 - 27 | 3065 | 68 | ||
| HQ529471.1 | - | 25 - 26 | 2972 | 68 | ||
| HQ529473.1 | - | 26 - 27 | 3146 | 68 | ||
| HQ529475.1 | - | 27 - 28 | 3158 | 68 | ||
| HQ529477.1 | - | 25 - 26 | 2992 | 68 | ||
| L16912.1 | - | 26 - 27 | 3063 | 68 | ||
| U09383.1 | - | 27 - 28 | 4006 | 68 | ||
Set Membership: |
|