Search
Search
| Gene Symbol: Cog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013581.3 | NP_038609.3 | 3 - 4 | 770 | 75 | |
| GenBank mRNA | AF109377.2 | - | 3 - 4 | 753 | 75 | |
| AK031510.1 | - | 2 - 3 | 226 | 75 | ||
| AK034636.2 | - | 3 - 4 | 775 | 75 | ||
| AK152464.1 | - | 3 - 4 | 761 | 75 | ||
| AK158507.1 | - | 3 - 4 | 1132 | 75 | ||
| AK159497.1 | - | 3 - 4 | 772 | 75 | ||
| AK162971.1 | - | 3 - 4 | 771 | 75 | ||
| AK171574.1 | - | 3 - 4 | 764 | 75 | ||
| AK220387.1 | - | 3 - 4 | 767 | 75 | ||
| BC063056.1 | - | 3 - 4 | 752 | 5720460 | 75 | |
Set Membership: |
|