Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516733_m1
          See other COIL GE Assays ›
          Gene Symbol:
          Coil
          Gene Name:
          coilin
          Gene Aliases:
          C79982, Cln80, p80, p80-coilin
          Chromosome Location:
          Chr.11: 88973935 - 88991613 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516733_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coil
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016706.2 NP_057915.2 6 - 7 1662 69
          GenBank mRNA AA240407.1 - 4 - 5 262 69
          AF208663.1 - 6 - 7 1644 69
          AK146366.1 - 6 - 7 1646 69
          BC031167.1 - 6 - 7 1677 4983919 69
          BM946023.1 - 1 - 2 99 69
          CF105200.1 - 5 - 6 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001385.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coil

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          protein binding
          protein C-terminus binding
          disulfide oxidoreductase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093291 CK792602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093291 AK146366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093291 BC031167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093291 BX630395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2093291 NM_016706.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 577768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78444 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159548 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112560 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093291 CK792602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093291 AK146366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093291 BC031167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093291 BX630395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2093291 NM_016706.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483839 AK146366.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483839 BC031167.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483839 BX630395.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483839 AF208663.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159548 - -
          Whole Mouse Genome G4122A A_51_P483839 AK146366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483839 BC031167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483839 BX630395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483839 AF208663.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137341 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112560 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09726298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09064916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09726296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09557229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09037941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380258 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380257 - -
          Affymetrix HT MG-430 PM Array Plate 1416692_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380252 - -
          Mouse Genome 430 2.0 Array 1416692_at - -
          Mouse Genome 430 2.0 Array 1416692_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12812_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12812_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100012985.mm.1 NM_016706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012985.mm.1 NM_016706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012985.mm.1 TC1100001385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012986.mm.1 NM_016706.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012986.mm.1 NM_016706.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012986.mm.1 TC1100001385.mm.1 MTA TC
          Mouse Expression Set 430 1416692_at - -
          Mouse Expression Set 430 1416692_PM_at - -
          Murine Genome U74v2 Set 97377_at - -
          Murine Genome U74v2 Set 97377_PM_at - -
          Mouse Genome 430A 2.0 Array 1416692_at - -
          Mouse Genome 430A 2.0 Array 1416692_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416692_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416692_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339631 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4480408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746210 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746210 - -
          mouse-6 v1.1 (Illumina) scl41054.7_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41054.7_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2746210 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3096250 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746210 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3096250 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41054.7_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline