Search
Search

Fanca| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016925.3 | NP_058621.2 | 6 - 7 | 606 | 56 | |
| GenBank mRNA | AF178934.1 | - | 6 - 7 | 602 | 56 | |
| AF208116.1 | - | 6 - 7 | 608 | 56 | ||
| AF208117.1 | - | 3 - 4 | 289 | 56 | ||
| AF208118.1 | - | 6 - 7 | 608 | 56 | ||
| AK084990.1 | - | 5 - 6 | 714 | 56 | ||
| AK166112.1 | - | 6 - 7 | 591 | 56 | ||
| BC150705.1 | - | 6 - 7 | 604 | 9087616 | 56 | |
| CF537383.1 | - | 6 - 7 | 523 | 56 | ||
Set Membership: |
|