Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517638_g1
          See other CLDN15 GE Assays ›
          Gene Symbol:
          Cldn15
          Gene Name:
          claudin 15
          Gene Aliases:
          2210009B08Rik, BB107105
          Chromosome Location:
          Chr.5: 136967869 - 136975845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517638_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cldn15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021719.4 NP_068365.1 4 - 5 817 112
          GenBank mRNA AK008227.1 - 4 - 5 615 112
          AK008683.1 - 4 - 5 818 112
          BC023428.1 - 4 - 5 806 5250004 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001614.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cldn15

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 AK008683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 BB801129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 BC023428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 AK008683.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 BB801129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 BC023428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355329 AK034724.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102362 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102578 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 AK008683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 BB801129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 BC023428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 NM_021719.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 AK008683.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 BB801129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 BC023428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 AK008683.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 BB801129.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 BC023428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 AK008683.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 BB801129.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 BC023428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24123909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01231111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102578 - -
          Whole Mouse Genome G4122A A_51_P306710 AK008683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 BB801129.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 BC023428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 NM_021719.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 AK008683.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 BB801129.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 BC023428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306710 NM_021719.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129367 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04982082 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04982090 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04982114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04982083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04982096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24123957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04908686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04725180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04908691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24123915 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526517 - -
          Affymetrix HT MG-430 PM Array Plate 1418920_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526514 - -
          Mouse Genome 430 2.0 Array 1418920_PM_at - -
          Mouse Genome 430 2.0 Array 1418920_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60363_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60363_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500014592.mm.1 NM_021719.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014592.mm.1 NM_021719.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014592.mm.1 TC0500001614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014593.mm.1 NM_021719.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014593.mm.1 NM_021719.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014593.mm.1 TC0500001614.mm.1 MTA TC
          Mouse Expression Set 430 1418920_at - -
          Mouse Expression Set 430 1418920_PM_at - -
          Murine Genome U74v2 Set 167191_at - -
          Murine Genome U74v2 Set 167191_PM_at - -
          Mouse Genome 430A 2.0 Array 1418920_PM_at - -
          Mouse Genome 430A 2.0 Array 1418920_at - -
          Affymetrix HT Mouse Genome 430A Array 1418920_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418920_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4730592 - -
          Illumina mouse-6 v1.0 expression beadchip 4150270 - -
          Illumina mouse-6 v1.0 expression beadchip 4730592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674906 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245622 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674906 - -
          mouse-6 v1.1 (Illumina) scl0004097.1_46-S - -
          mouse-6 v1.1 (Illumina) scl27110.7.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150270 - -
          Illumina Sentrix mouse-6.v1 bead array 4730592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27110.7.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004097.1_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674906 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245622 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793817 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674906 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793817 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline