Search
Search

Impg1| Gene Symbol: Impg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022016.3 | NP_071299.3 | 4 - 5 | 643 | 95 | |
| GenBank mRNA | AF229929.1 | - | 4 - 5 | 554 | 95 | |
| AF266478.1 | - | 4 - 5 | 689 | 95 | ||
| AK020862.1 | - | 3 - 4 | 413 | 95 | ||
| BC022970.1 | - | 4 - 5 | 631 | 5366008 | 95 | |
Set Membership: |
|