Search
Search

Hspa5| Gene Symbol: Hspa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163434.1 | NP_001156906.1 | 5 - 6 | 762 | 72 | |
| NM_022310.3 | NP_071705.3 | 4 - 5 | 844 | 72 | ||
| GenBank mRNA | AJ002387.1 | - | 4 - 5 | 623 | 72 | |
| AK004578.1 | - | 4 - 5 | 839 | 72 | ||
| AK076079.1 | - | 5 - 6 | 758 | 72 | ||
| AK077463.1 | - | 5 - 6 | 762 | 72 | ||
| AK144650.1 | - | 5 - 6 | 758 | 72 | ||
| AK144742.1 | - | 5 - 6 | 758 | 72 | ||
| AK146647.1 | - | 5 - 6 | 761 | 72 | ||
| AK148539.1 | - | 5 - 6 | 761 | 72 | ||
| AK149447.1 | - | 5 - 6 | 761 | 72 | ||
| AK151244.1 | - | 4 - 5 | 711 | 72 | ||
| AK151647.1 | - | 5 - 6 | 760 | 72 | ||
| AK151806.1 | - | 5 - 6 | 761 | 72 | ||
| AK152020.1 | - | 5 - 6 | 758 | 72 | ||
| AK152521.1 | - | 4 - 5 | 841 | 72 | ||
| AK152958.1 | - | 5 - 6 | 761 | 72 | ||
| AK159719.1 | - | 5 - 6 | 758 | 72 | ||
| AK160821.1 | - | 761 | 72 | |||
| AK166207.1 | - | 5 - 6 | 758 | 72 | ||
| AK166739.1 | - | 5 - 6 | 758 | 72 | ||
| AK167011.1 | - | 5 - 6 | 758 | 72 | ||
| AK168012.1 | - | 5 - 6 | 762 | 72 | ||
| AK169034.1 | - | 5 - 6 | 762 | 72 | ||
| AK192496.1 | - | 3 - 4 | 353 | 72 | ||
| AK194376.1 | - | 3 - 4 | 72 | |||
| AK195133.1 | - | 3 - 4 | 318 | 72 | ||
| AK197130.1 | - | 3 - 4 | 309 | 72 | ||
| AK198934.1 | - | 3 - 4 | 317 | 72 | ||
| AK200033.1 | - | 3 - 4 | 316 | 72 | ||
| BC005785.1 | - | 3 - 4 | 478 | 72 | ||
| BC050927.1 | - | 4 - 5 | 821 | 6415084 | 72 | |
| D78645.1 | - | 4 - 5 | 704 | 72 | ||
Set Membership: |
|