Search
Search

Irak3| Gene Symbol: Irak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028679.3 | NP_082955.2 | 2 - 3 | 396 | 68 | |
| GenBank mRNA | AF461763.1 | - | 2 - 3 | 376 | 68 | |
| AJ440757.2 | - | 2 - 3 | 318 | 68 | ||
| AK014783.1 | - | 2 - 3 | 447 | 68 | ||
| AK028436.1 | - | 309 | 68 | |||
| AK029057.1 | - | 2 - 3 | 397 | 68 | ||
| AK049210.1 | - | 2 - 3 | 427 | 68 | ||
| BC120829.1 | - | 2 - 3 | 357 | 40129752 | 68 | |
| BC120831.1 | - | 2 - 3 | 357 | 40129754 | 68 | |
| BC144835.1 | - | 2 - 3 | 357 | 68 | ||
Set Membership: |
|