Search
Search
| Gene Symbol: Cul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029402.3 | NP_083678.2 | 13 - 14 | 1496 | 119 | |
| GenBank mRNA | AK004875.2 | - | 13 - 14 | 1452 | 119 | |
| AK016520.1 | - | 13 - 14 | 1466 | 119 | ||
| AK028589.1 | - | 13 - 14 | 1465 | 119 | ||
| AK144693.1 | - | 13 - 14 | 1470 | 119 | ||
| AK160597.1 | - | 13 - 14 | 1481 | 119 | ||
| AK220287.1 | - | 13 - 14 | 1471 | 119 | ||
| BC025902.1 | - | 5 - 6 | 562 | 119 | ||
| BC026779.1 | - | 13 - 14 | 1397 | 4018298 | 119 | |
| BC027428.1 | - | 13 - 14 | 1459 | 3983461 | 119 | |
Set Membership: |
|