Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Creb3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB047647.1 | - | 3 - 4 | 393 | 89 | |
AB052779.1 | - | 5 - 6 | 650 | 89 | ||
AB182651.1 | - | 5 - 6 | 768 | 89 | ||
AB182652.1 | - | 3 - 4 | 393 | 89 | ||
AF287260.1 | - | 3 - 4 | 531 | 89 | ||
AK005918.1 | - | 3 - 4 | 550 | 89 | ||
AK019928.1 | - | 5 - 6 | 768 | 89 | ||
AK088919.1 | - | 5 - 6 | 667 | 89 | ||
AY049978.1 | - | 5 - 6 | 689 | 89 | ||
BC022605.1 | - | 5 - 6 | 624 | 4164994 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |