Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053208.4 | NP_444438.2 | 4 - 5 | 1481 | 71 | |
GenBank mRNA | AF340231.1 | - | 3 - 4 | 1223 | 71 | |
AF453879.1 | - | 4 - 5 | 1403 | 71 | ||
AJ310547.1 | - | 4 - 5 | 1452 | 71 | ||
AK075582.1 | - | 4 - 5 | 1433 | 71 | ||
AK152056.1 | - | 4 - 5 | 1442 | 71 | ||
AK152223.1 | - | 4 - 5 | 1439 | 71 | ||
AK159206.1 | - | 4 - 5 | 1434 | 71 | ||
AK166661.1 | - | 5 - 6 | 1347 | 71 | ||
AK167477.1 | - | 4 - 5 | 1434 | 71 | ||
AK179611.1 | - | 1 - 2 | 62 | 71 | ||
BC023299.1 | - | 4 - 5 | 1415 | 5135579 | 71 | |
BC051436.1 | - | 3 - 4 | 1199 | 6312638 | 71 | |
BC086764.1 | - | 4 - 5 | 1420 | 5685311 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |