Search
Search

Cacng7| Gene Symbol: Cacng7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133189.3 | NP_573452.3 | 2 - 3 | 281 | 76 | |
| GenBank mRNA | AF361349.1 | - | 2 - 3 | 281 | 76 | |
| AF458899.1 | - | 2 - 3 | 281 | 76 | ||
| AK032173.1 | - | 3 - 4 | 393 | 76 | ||
| BC141277.1 | - | 2 - 3 | 319 | 9055889 | 76 | |
Set Membership: |
|