Search
Search

Agap3| Gene Symbol: Agap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139153.2 | NP_631892.2 | 14 - 15 | 1988 | 70 | |
| GenBank mRNA | AF459091.1 | - | 14 - 15 | 1988 | 70 | |
| AK079516.1 | - | 10 - 11 | 1327 | 70 | ||
| BC031173.1 | - | 5 - 6 | 560 | 70 | ||
| BM943884.1 | - | 1 - 2 | 53 | 70 | ||
Set Membership: |
|