Search
Search

Dlgap5| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 9 - 10 | 1422 | 108 | |
| GenBank mRNA | AB076696.1 | - | 9 - 10 | 1422 | 108 | |
| AK036384.1 | - | 9 - 10 | 1224 | 108 | ||
| AK048470.1 | - | 9 - 10 | 1192 | 108 | ||
| AK085882.1 | - | 9 - 10 | 1203 | 108 | ||
| AK129032.1 | - | 9 - 10 | 1168 | 108 | ||
| AK135820.1 | - | 9 - 10 | 1229 | 108 | ||
| AK147955.1 | - | 9 - 10 | 1192 | 108 | ||
| BC043924.1 | - | 1 - 2 | 48 | 108 | ||
| BC058087.1 | - | 9 - 10 | 1156 | 6816660 | 108 | |
Set Membership: |
|