Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145148.2 | NP_660130.2 | 10 - 11 | 1156 | 104 | |
GenBank mRNA | AB042027.1 | - | 10 - 11 | 1079 | 104 | |
AF327856.1 | - | 9 - 10 | 779 | 104 | ||
AF327857.1 | - | 8 - 9 | 566 | 104 | ||
AK129262.1 | - | 9 - 10 | 631 | 104 | ||
AK143978.1 | - | 9 - 10 | 778 | 104 | ||
BC052390.1 | - | 10 - 11 | 1025 | 5718942 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |