| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500004449.mm.1 |
NM_145471.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004449.mm.1 |
NM_145471.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004449.mm.1 |
TC1500000607.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500004445.mm.1 |
NM_145471.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004445.mm.1 |
NM_145471.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004445.mm.1 |
TC1500000607.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500004448.mm.1 |
NM_145471.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004448.mm.1 |
NM_145471.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500004448.mm.1 |
TC1500000607.mm.1 |
MTA TC |