Search
Search

Pprc1| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081214.1 | NP_001074683.1 | 12 - 13 | 4732 | 105 | |
| GenBank mRNA | AK122322.1 | - | 12 - 13 | 4687 | 105 | |
| AK171995.1 | - | 7 - 8 | 3325 | 105 | ||
| BC013720.1 | - | 8 - 9 | 2099 | 105 | ||
| BC066048.1 | - | 12 - 13 | 4686 | 6825158 | 105 | |
Set Membership: |
|