Search
Search
| Gene Symbol: Lias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024471.4 | NP_077791.1 | 2 - 3 | 347 | 96 | |
| GenBank mRNA | AB057731.1 | - | 2 - 3 | 246 | 96 | |
| AK031851.1 | - | 2 - 3 | 319 | 96 | ||
| AK048744.1 | - | 291 | 96 | |||
| AK049548.1 | - | 2 - 3 | 262 | 96 | ||
| AK077192.1 | - | 2 - 3 | 348 | 96 | ||
| BC002141.1 | - | 2 - 3 | 279 | 3484559 | 96 | |
Set Membership: |
|