Search
Search
| Gene Symbol: Utp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144826.3 | NP_659075.1 | 7 - 8 | 625 | 67 | |
| GenBank mRNA | AF334387.1 | - | 7 - 8 | 952 | 67 | |
| AK029151.1 | - | 7 - 8 | 626 | 67 | ||
| AK084976.1 | - | 7 - 8 | 618 | 67 | ||
| AK169785.1 | - | 7 - 8 | 632 | 67 | ||
| BC018250.1 | - | 7 - 8 | 589 | 4924739 | 67 | |
| BC023920.1 | - | 7 - 8 | 619 | 5323367 | 67 | |
| BC024850.1 | - | 7 - 8 | 620 | 5360921 | 67 | |
Set Membership: |
|