Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00522856_m1
          See other SERPINA10 GE Assays ›
          Gene Symbol:
          Serpina10
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
          Gene Aliases:
          PZI, ZPI
          Chromosome Location:
          Chr.12: 103616675 - 103631490 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00522856_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301404.1 NP_001288333.1 4 - 5 1091 58
          NM_144834.4 NP_659083.2 4 - 5 1253 58
          GenBank mRNA AK050552.1 - 1230 58
          AY324633.1 - 4 - 5 1253 58
          BC018416.1 - 4 - 5 1068 4196269 58
          BC025821.1 - 3 - 4 1215 5100400 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002271.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpina10

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          hemostasis
          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          serine-type endopeptidase inhibitor activity
          heparin binding
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010196 BC025821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010196 AK050552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010196 AY324633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010196 NM_144834.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010197 BC025821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010197 AK050552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010197 AY324633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010197 BC018416.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010197 NM_144834.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558963 AK136731.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558963 BC028789.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54961 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139445 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160582 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010197 BC025821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010197 AK050552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010197 AY324633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010197 BC018416.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010197 NM_144834.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010196 BC025821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010196 AK050552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010196 AY324633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010196 NM_144834.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28698619 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02594401 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160582 - -
          Whole Mouse Genome G4122A A_51_P472033 AK050552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662025 BC025821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662025 AK050552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662025 AY324633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662025 BC018416.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662025 NM_144834.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114150 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160582 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139445 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138952 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10216460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10216452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10216451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10216462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10138951 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09964874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28698683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10138951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10433785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09864331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09964874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09845219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09864341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10138952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09864347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10412397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10423077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10423074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28698627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10138944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402367 - -
          Affymetrix HT MG-430 PM Array Plate 1424758_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402360 - -
          Mouse Genome 430 2.0 Array 1424758_s_at - -
          Mouse Genome 430 2.0 Array 1424758_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217847_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217847_at - -
          Affymetrix Mouse Clariom S Array PSR1200015771.mm.1 NM_144834.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015771.mm.1 TC1200002271.mm.1 MTA TC
          Mouse Expression Set 430 1424758_PM_s_at - -
          Mouse Expression Set 430 1424758_s_at - -
          Murine Genome U74v2 Set 96787_at - -
          Murine Genome U74v2 Set 96787_PM_at - -
          Mouse Genome 430A 2.0 Array 1424758_s_at - -
          Mouse Genome 430A 2.0 Array 1424758_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424758_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424758_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 666012 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2360114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259215 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259215 - -
          mouse-6 v1.1 (Illumina) scl0217847.1_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0217847.1_55-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259215 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline