Search
Search

Klhl20| Gene Symbol: Klhl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039482.1 | NP_001034571.1 | 2 - 3 | 180 | 58 | |
| GenBank mRNA | AK053102.1 | - | 2 - 3 | 130 | 58 | |
| AK053371.1 | - | 2 - 3 | 144 | 58 | ||
| AK054169.1 | - | 2 - 3 | 148 | 58 | ||
| AK220547.1 | - | 2 - 3 | 159 | 58 | ||
| BC019571.1 | - | 2 - 3 | 152 | 4235202 | 58 | |
| BC060183.1 | - | 2 - 3 | 109 | 58 | ||
Set Membership: |
|