Search
Search
| Gene Symbol: Mavs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206385.1 | NP_001193314.1 | 2 - 3 | 242 | 60 | |
| NM_144888.2 | NP_659137.1 | 2 - 3 | 470 | 60 | ||
| GenBank mRNA | AK028421.1 | - | 2 - 3 | 241 | 60 | |
| AK085038.1 | - | 2 - 3 | 429 | 60 | ||
| AX775773.1 | - | 2 - 3 | 219 | 60 | ||
| BC020006.1 | - | 2 - 3 | 202 | 4190175 | 60 | |
| BC025825.1 | - | 2 - 3 | 395 | 5100990 | 60 | |
| BC031352.1 | - | 2 - 3 | 379 | 5123637 | 60 | |
| BC037391.1 | - | 1 - 2 | 181 | 5361522 | 60 | |
| DQ167127.1 | - | 1 - 2 | 119 | 60 | ||
| DQ174271.1 | - | 1 - 2 | 119 | 60 | ||
Set Membership: |
|