Search
Search

Ndufs1| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 15 - 16 | 2059 | 70 | |
| NM_001160039.1 | NP_001153511.1 | 15 - 16 | 1901 | 70 | ||
| NM_001160040.1 | NP_001153512.1 | 14 - 15 | 1955 | 70 | ||
| NM_145518.2 | NP_663493.2 | 14 - 15 | 1797 | 70 | ||
| GenBank mRNA | AK034597.1 | - | 14 - 15 | 1866 | 70 | |
| AK036926.1 | - | 14 - 15 | 1662 | 70 | ||
| AK142711.1 | - | 14 - 15 | 1677 | 70 | ||
| AK146814.1 | - | 14 - 15 | 1815 | 70 | ||
| AK167705.1 | - | 14 - 15 | 1645 | 70 | ||
| AK196215.1 | - | 1 - 2 | 44 | 70 | ||
| AK219412.1 | - | 1 - 2 | 44 | 70 | ||
| BC006660.1 | - | 14 - 15 | 1684 | 3501043 | 70 | |
| BC015300.1 | - | 14 - 15 | 1617 | 4236882 | 70 | |
Set Membership: |
|