Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524497_m1
          See other BUD13 GE Assays ›
          Gene Symbol:
          Bud13
          Gene Name:
          BUD13 homolog (yeast)
          Gene Aliases:
          AV305342, D030060M11Rik
          Chromosome Location:
          Chr.9: 46283012 - 46298783 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524497_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bud13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146000.2 NP_666112.1 5 - 6 1326 121
          GenBank mRNA AK037134.1 - 5 - 6 1323 121
          AK038126.1 - 5 - 6 1331 121
          AK051047.1 - 5 - 6 1331 121
          AK077744.1 - 5 - 6 1330 121
          BC025490.1 - 5 - 6 1327 5252666 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000563.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bud13

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478982 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 BC025490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 NM_146000.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 AK038126.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 BC025490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 NM_146000.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159284 AK038126.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53654 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119111 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153156 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 BC025490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 NM_146000.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 AK038126.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 BC025490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 NM_146000.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159284 AK038126.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478987 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153156 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119111 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06774619 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06774630 - -
          Whole Mouse Genome G4122A A_52_P435394 AK051047.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 BC025490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 NM_146000.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 AK038126.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 BC025490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 NM_146000.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159284 AK038126.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123935 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153156 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119111 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08143909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08143917 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26478982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26479063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07516894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07622361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08004983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07590195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07516897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07596029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07590197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07725334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07516896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08068190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26479102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585043 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585044 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585041 - -
          Affymetrix HT MG-430 PM Array Plate 1460355_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585037 - -
          Mouse Genome 430 2.0 Array 1460355_at - -
          Mouse Genome 430 2.0 Array 1460355_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215051_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215051_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900003983.mm.1 NM_146000.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003983.mm.1 NM_146000.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003983.mm.1 TC0900000563.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900003984.mm.1 NM_146000.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003984.mm.1 NM_146000.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003984.mm.1 TC0900000563.mm.1 MTA TC
          Mouse Expression Set 430 1460355_PM_at - -
          Mouse Expression Set 430 1460355_at - -
          Murine Genome U74v2 Set 170775_PM_at - -
          Murine Genome U74v2 Set 170775_at - -
          Mouse Genome 430A 2.0 Array 1460355_at - -
          Mouse Genome 430A 2.0 Array 1460355_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460355_at - -
          Affymetrix HT Mouse Genome 430A Array 1460355_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 389768 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6220097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593532 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2593532 - -
          mouse-6 v1.1 (Illumina) scl36998.11.1_88-S - -
          Illumina Sentrix mouse-6.v1 bead array 6220097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36998.11.1_88-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2593532 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593532 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36998.11.1_88-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline