Search
Search
| Gene Symbol: Ccm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190343.1 | NP_001177272.1 | 4 - 5 | 546 | 76 | |
| NM_001190344.1 | NP_001177273.1 | 5 - 6 | 632 | 76 | ||
| NM_146014.3 | NP_666126.1 | 5 - 6 | 720 | 76 | ||
| GenBank mRNA | BC029157.1 | - | 5 - 6 | 651 | 4952288 | 76 |
Set Membership: |
|