Search
Search
| Gene Symbol: Eml6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146016.2 | NP_666128.2 | 38 - 39 | 6161 | 70 | |
| GenBank mRNA | AK081418.1 | - | 3 - 4 | 512 | 70 | |
| BC024726.1 | - | 10 - 11 | 1596 | 5362808 | 70 | |
Set Membership: |
|