Search
Search

Golph3l| Gene Symbol: Golph3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177670.1 | NP_001171141.1 | 2 - 3 | 391 | 103 | |
| NM_146133.3 | NP_666245.2 | 2 - 3 | 391 | 103 | ||
| GenBank mRNA | AK167675.1 | - | 2 - 3 | 341 | 103 | |
| AK169724.1 | - | 2 - 3 | 345 | 103 | ||
| BC027194.1 | - | 2 - 3 | 301 | 4216692 | 103 | |
| BC047147.1 | - | 2 - 3 | 366 | 6465983 | 103 | |
Set Membership: |
|