Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00526114_s1
          See other OLFR958 GE Assays ›
          Gene Symbol:
          Olfr958
          Gene Name:
          olfactory receptor 958
          Gene Aliases:
          MOR224-9
          Chromosome Location:
          Chr.9: 39549931 - 39550869 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00526114_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr958
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146330.1 NP_666442.1 1 147 68
          GenBank mRNA BC148230.1 - 1 151 40142225 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002081.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr958

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132101 NM_146330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132101 NM_146330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149752 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132101 NM_146330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132101 BC148230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132101 NM_146330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132101 NM_146330.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132101 NM_146330.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26439005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149752 - -
          Whole Mouse Genome G4122A A_51_P132101 NM_146330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132101 BC148230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132101 NM_146330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P132101 BC148230.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105118 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149752 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156900 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156910 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26439019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07762506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08156901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08156902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07913367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07550960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07886055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08091332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26439008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592415 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592414 - -
          Affymetrix Mouse Clariom S Array PSR0900017210.mm.1 NM_146330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017210.mm.1 NM_146330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017210.mm.1 TC0900002081.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 551508 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6370358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233657 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233657 - -
          mouse-6 v1.1 (Illumina) scl36010.1.352_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36010.1.352_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233657 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233657 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline