Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00528142_s1
          See other OLFR466 GE Assays ›
          Gene Symbol:
          Olfr466
          Gene Name:
          olfactory receptor 466
          Gene Aliases:
          MOR209-1
          Chromosome Location:
          Chr.13: 65152226 - 65153152 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00528142_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr466
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146819.2 NP_667030.2 1 448 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000787.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr466

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174851 NM_146819.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155791 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174851 NM_146819.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P133456 NM_146819.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102029 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155791 - -
          Whole Mouse Genome G4122A A_51_P133456 NM_146819.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134408 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10682208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10928433 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10682209 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10928430 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10928441 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10928436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29101997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10761229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10928428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10928433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10928439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11005831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10630880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10495587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29101997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10718486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29101998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405850 - -
          Affymetrix Mouse Gene 1.1 ST Array 10405849 - -
          Affymetrix Mouse Clariom S Array PSR1300005197.mm.1 NM_146819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005197.mm.1 NM_146819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300005197.mm.1 TC1300000787.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 792049 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320070 - -
          Illumina mouse-6 v1.0 expression beadchip 2320070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769858 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769858 - -
          mouse-6 v1.1 (Illumina) scl12188.1.1_113-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl12188.1.1_113-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2769858 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline