Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00529886_s1
          See other OLFR652 GE Assays ›
          Gene Symbol:
          Olfr652
          Gene Name:
          olfactory receptor 652
          Gene Aliases:
          MOR318
          Chromosome Location:
          Chr.7: 104564152 - 104565212 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00529886_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr652
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147048.2 NP_667259.2 1 531 87
          GenBank mRNA BC137763.1 - 1 531 8860782 87
          BC137764.1 - 1 531 8860783 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001524.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr652

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962655 NM_147048.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962655 BC137763.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962655 BC137764.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105197 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962655 NM_147048.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962655 BC137763.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962655 BC137764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203943 NM_147048.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203943 BC137763.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203943 BC137764.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105197 - -
          Whole Mouse Genome G4122A A_51_P203943 NM_147048.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203943 BC137763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203943 BC137764.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100885 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123853 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06036762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06518305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06287536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06512260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06512263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06209330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06518300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388558 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555874 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555873 - -
          Affymetrix Mouse Clariom S Array PSR0700012891.mm.1 TC0700001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012891.mm.1 NM_147048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012891.mm.1 NM_147048.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012890.mm.1 TC0700001524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012890.mm.1 NM_147048.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012890.mm.1 NM_147048.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 578756 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233010 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233010 - -
          mouse-6 v1.1 (Illumina) scl32254.1.1_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32254.1.1_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233010 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline