Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00530473_s1
          See other OLFR644 GE Assays ›
          Gene Symbol:
          Olfr644 Olfr643
          Gene Name:
          olfactory receptor 644
          olfactory receptor 643
          Gene Aliases:
          MOR13-1
          MOR13-2
          Chromosome Location:
          Chr.7: 104068085 - 104069029 on Build GRCm38
          Chromosome Location:
          Chr.7: 104058619 - 104137416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00530473_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr644
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147121.1 NP_667332.1 1 465 78
          GenBank mRNA BC099887.1 - 1 457 78
          Gene Symbol: Olfr643
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147077.1 NP_667288.1 1 78
          GenBank mRNA BC106803.2 - 1 40042567 78
          BC146607.1 - 1 40042594 78
          BC148182.1 - 1 40043246 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003921.mm.1, TC0700003924.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr644
          Click here to find siRNAs for Gene Olfr643

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 NM_147121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 BC099886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 BC099887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 NM_147121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 BC099886.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413886 BC099887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 NM_147121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 BC099886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 BC099887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 NM_147121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 BC099886.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413886 BC099887.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 NM_147121.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 BC099886.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 BC099887.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 NM_147121.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 BC099886.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413886 BC099887.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386497 - -
          Whole Mouse Genome G4122A A_51_P413886 NM_147121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413886 BC099886.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413886 BC099887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413886 NM_147121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413886 BC099886.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413886 BC099887.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06225335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06225325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06225335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386537 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566301 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566300 - -
          Affymetrix Mouse Clariom S Array PSR0700032828.mm.1 NM_147077.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032828.mm.1 NM_147077.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032828.mm.1 TC0700003921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032835.mm.1 NM_147121.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032835.mm.1 NM_147121.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032835.mm.1 TC0700003924.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 385303 - -
          ABI Mouse Genome Survey Microarray 622685 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1580142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717767 - -
          mouse-6 v1.1 (Illumina) scl0259125.1_241-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0259125.1_241-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717767 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline