Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pop4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025390.4 | NP_079666.1 | 2 - 3 | 256 | 116 | |
GenBank mRNA | AK002855.1 | - | 2 - 3 | 257 | 116 | |
AK003293.2 | - | 2 - 3 | 257 | 116 | ||
AK003908.1 | - | 2 - 3 | 271 | 116 | ||
AK054466.1 | - | 288 | 116 | |||
AK150459.1 | - | 2 - 3 | 263 | 116 | ||
AK152948.1 | - | 2 - 3 | 263 | 116 | ||
BC011465.1 | - | 2 - 3 | 272 | 3966371 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |