Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546658_s1
          See other CDC42EP2 GE Assays ›
          Gene Symbol:
          Cdc42ep2
          Gene Name:
          CDC42 effector protein (Rho GTPase binding) 2
          Gene Aliases:
          1110008C05Rik, Borg1, Cep2, D19Bwg1013e
          Chromosome Location:
          Chr.19: 5917556 - 5924816 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546658_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc42ep2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026772.2 NP_081048.1 2 998 82
          GenBank mRNA AK003562.1 - 1 173 82
          AK083004.1 - 1000 82
          BC034884.1 - 1 956 5257253 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc42ep2

          Gene Ontology Categories:

          Function(s) Process(es)

          Rho protein signal transduction
          regulation of cell shape
          actin cytoskeleton organization
          positive regulation of actin filament polymerization
          positive regulation of pseudopodium assembly
          positive regulation of GTPase activity
          opioid peptide activity
          GTPase activator activity
          GTP-Rho binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 AK083004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 BC034884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 NM_026772.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 AK003562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 AK083004.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 BC034884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 NM_026772.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155554 AK003562.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632828 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174460 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 AK083004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 BC034884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 NM_026772.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 AK003562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 AK083004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 BC034884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 NM_026772.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155554 AK003562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 AK083004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 BC034884.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 NM_026772.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 AK003562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 AK083004.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 BC034884.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 NM_026772.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P155554 AK003562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230214 AK083004.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31931687 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09094844 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174460 - -
          Whole Mouse Genome G4122A A_52_P155554 AK083004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 BC034884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 NM_026772.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 AK003562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 AK083004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 BC034884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 NM_026772.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P155554 AK003562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230214 AK083004.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124981 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14342765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31931712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14487084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14389490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14290192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14153927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14326684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14219943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14153926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14376919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14342762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14153935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31931713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14290187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465280 - -
          Affymetrix HT MG-430 PM Array Plate 1428750_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465278 - -
          Mouse Genome 430 2.0 Array 1428750_at - -
          Mouse Genome 430 2.0 Array 1428750_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104252_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104252_at - -
          Mouse Expression Set 430 1428750_PM_at - -
          Mouse Expression Set 430 1428750_at - -
          Affymetrix HT Mouse Genome 430B Array 1428750_at - -
          Affymetrix HT Mouse Genome 430B Array 1428750_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 584309 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105670465 - -
          Illumina mouse-6 v1.0 expression beadchip 105670465 - -
          Illumina mouse-6 v1.0 expression beadchip 540079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239294 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251299 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239294 - -
          mouse-6 v1.1 (Illumina) scl00104252.2_200-S - -
          mouse-6 v1.1 (Illumina) scl53518.2_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 105670465 - -
          Illumina Sentrix mouse-6.v1 bead array 540079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00104252.2_200-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105670465 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239294 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239294 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00104252.2_200-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline