| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700021071.mm.1 |
NM_027292.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021071.mm.1 |
NM_027292.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021071.mm.1 |
TC1700002227.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700021070.mm.1 |
NM_027292.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021070.mm.1 |
NM_027292.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021070.mm.1 |
TC1700002227.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700021072.mm.1 |
NM_027292.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021072.mm.1 |
NM_027292.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700021072.mm.1 |
TC1700002227.mm.1 |
MTA TC |