Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546933_m1
          See other 4632404H12RIK GE Assays ›
          Gene Symbol:
          4632404H12Rik
          Gene Name:
          RIKEN cDNA 4632404H12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 89768430 - 89773233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546933_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4632404H12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK014549.2 - 741 84
          BC106127.1 - 1 - 2 745 84

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK014549.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321611 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483483 AK014549.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483483 AK014549.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22321606 - -
          Whole Mouse Genome G4122A A_52_P389863 AK164170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389863 AK169469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389863 AW045319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389863 BC063049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P389863 AK048101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483483 AK014549.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483483 AK014549.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133135 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22321608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499653 - -
          Affymetrix HT MG-430 PM Array Plate 1438209_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431820_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499652 - -
          Mouse Genome 430 2.0 Array 1438209_at - -
          Mouse Genome 430 2.0 Array 1438209_PM_at - -
          Mouse Genome 430 2.0 Array 1431820_at - -
          Mouse Genome 430 2.0 Array 1431820_PM_at - -
          Mouse Expression Set 430 1431820_at - -
          Mouse Expression Set 430 1438209_at - -
          Mouse Expression Set 430 1438209_PM_at - -
          Mouse Expression Set 430 1431820_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438209_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431820_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438209_at - -
          Affymetrix HT Mouse Genome 430B Array 1431820_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 757085 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5900168 - -
          Illumina mouse-6 v1.0 expression beadchip 5720484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005900168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629897 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629897 - -
          mouse-6 v1.1 (Illumina) scl0074034.1_260-S - -
          mouse-6 v1.1 (Illumina) scl074034.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 5900168 - -
          Illumina Sentrix mouse-6.v1 bead array 5720484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl074034.1_57-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0074034.1_260-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720484 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5900168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629897 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594873 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594872 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594872 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594873 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629897 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline