Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546961_m1
          See other NUAK2 GE Assays ›
          Gene Symbol:
          Nuak2
          Gene Name:
          NUAK family, SNF1-like kinase, 2
          Gene Aliases:
          1200013B22Rik, Omphk2, Snark, mKIAA0537
          Chromosome Location:
          Chr.1: 132316125 - 132333488 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546961_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nuak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195025.1 NP_001181954.1 1 - 2 353 97
          NM_028778.4 NP_083054.1 1 - 2 353 97
          GenBank mRNA AK004737.2 - 1 - 2 341 97
          AK033672.1 - 1 - 2 340 97
          AK034082.1 - 1 - 2 345 97
          BC033302.1 - 1 - 2 3663595 97
          BC046833.1 - 1 - 2 328 6816569 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001190.mm.1
          This assay targets the same location as RTA transcript cluster(s) TCUn_KL568212v100000001.rn

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nuak2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          phosphorylation
          actin cytoskeleton organization
          negative regulation of TOR signaling
          intracellular signal transduction
          cellular response to glucose starvation
          negative regulation of apoptotic process
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100009672.mm.1 NM_001195025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009672.mm.1 NM_001195025.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009672.mm.1 NM_028778.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009672.mm.1 NM_028778.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009672.mm.1 TC0100001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009679.mm.1 NM_001195025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009679.mm.1 NM_001195025.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009679.mm.1 NM_028778.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009679.mm.1 NM_028778.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009679.mm.1 TC0100001190.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009675.mm.1 NM_001195025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009675.mm.1 NM_001195025.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009675.mm.1 NM_028778.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009675.mm.1 NM_028778.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009675.mm.1 TC0100001190.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910820 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline