Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00547308_s1
          See other GM14458 GE Assays ›
          Gene Symbol:
          Gm14458 Fthl17 Gm14511 Gm5634 Gm5635
          Gene Name:
          predicted gene 14458
          ferritin, heavy polypeptide-like 17
          predicted gene 14511
          predicted gene 5634
          predicted gene 5635
          Gene Aliases:
          Fthl17L3, OTTMUSG00000016694
          -
          EG434727, Fthl17L2, OTTMUSG00000016933
          EG434726, Fthl17L1
          EG434729, Fthl17L5
          Chromosome Location:
          Chr.X: 8985940 - 8986672 on Build GRCm38
          Chromosome Location:
          Chr.X: 9033486 - 9034333 on Build GRCm38
          Chromosome Location:
          Chr.X: 8975718 - 8976559 on Build GRCm38
          Chromosome Location:
          Chr.X: 8962134 - 8962975 on Build GRCm38
          Chromosome Location:
          Chr.X: 9063090 - 9063822 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          Assay ID Mm00547308_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14458
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099326.2 NP_001092796.1 1 467 70
          GenBank mRNA BC104363.1 - 1 468 40049729 70
          Gene Symbol: Fthl17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031261.2 NP_112551.2 1 542 70
          GenBank mRNA AF285569.1 - 1 543 70
          Gene Symbol: Gm14511
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085525.2 NP_001078994.1 1 536 70
          GenBank mRNA CF106510.1 - 1 412 70
          Gene Symbol: Gm5634
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085524.1 NP_001078993.1 1 536 70
          Gene Symbol: Gm5635
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038697.2 NP_001033786.1 1 467 70
          GenBank mRNA BC104362.1 - 1 467 40049728 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm14458
          Click here to find siRNAs for Gene Fthl17
          Click here to find siRNAs for Gene Gm14511
          Click here to find siRNAs for Gene Gm5634
          Click here to find siRNAs for Gene Gm5635

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126848 NM_001038697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126848 BC104362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999883 NM_001085525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999883 NM_001038697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999883 BC104362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999882 NM_001099326.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999882 NM_001085524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999882 NM_001085525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1999882 AF285569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990271 NM_031261.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126848 NM_001038697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126848 BC104362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999883 NM_001085525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999883 NM_001038697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999883 BC104362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999882 NM_001099326.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999882 NM_001085524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999882 NM_001085525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1999882 AF285569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990271 NM_031261.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32245173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32244867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32245171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32245170 - -
          Whole Mouse Genome G4122A A_51_P223504 AF285569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P223504 NM_031261.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110753 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118083 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126848 NM_001038697.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126848 BC104362.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32245170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32244684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32245169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32245171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32245173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32244757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32244688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14771515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15185204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15185185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32244688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15072358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32244755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32244867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15185177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32244697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32245170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603544 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603544 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598565 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603543 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598567 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603543 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598569 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603545 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603545 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598565 - -
          Mouse Genome 430 2.0 Array 1419540_at - -
          Mouse Genome 430 2.0 Array 1419540_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000780.mm.1 NM_001038697.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000780.mm.1 TC0X00000104.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011684.mm.1 NM_001085524.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011684.mm.1 NM_001085524.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011684.mm.1 TC0X00001815.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00011687.mm.1 NM_001085525.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011687.mm.1 NM_001277184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011687.mm.1 TC0X00001816.mm.1 MTA TC
          Mouse Expression Set 430 1419540_PM_at - -
          Mouse Expression Set 430 1419540_PM_at - -
          Mouse Expression Set 430 1419540_at - -
          Mouse Expression Set 430 1419540_at - -
          Mouse Expression Set 430 1419540_at - -
          Mouse Expression Set 430 1419540_at - -
          Mouse Expression Set 430 1419540_PM_at - -
          Mouse Expression Set 430 1419540_PM_at - -
          Murine Genome U74v2 Set 168973_i_at - -
          Murine Genome U74v2 Set 168973_i_at - -
          Murine Genome U74v2 Set 170471_i_at - -
          Murine Genome U74v2 Set 170471_i_at - -
          Murine Genome U74v2 Set 170471_i_at - -
          Murine Genome U74v2 Set 168973_i_at - -
          Murine Genome U74v2 Set 168973_PM_i_at - -
          Murine Genome U74v2 Set 168973_PM_i_at - -
          Murine Genome U74v2 Set 170471_PM_i_at - -
          Murine Genome U74v2 Set 168973_i_at - -
          Murine Genome U74v2 Set 170471_PM_i_at - -
          Murine Genome U74v2 Set 168973_PM_i_at - -
          Murine Genome U74v2 Set 170471_i_at - -
          Murine Genome U74v2 Set 170471_i_at - -
          Murine Genome U74v2 Set 170471_PM_i_at - -
          Murine Genome U74v2 Set 170471_PM_i_at - -
          Murine Genome U74v2 Set 168973_PM_i_at - -
          Murine Genome U74v2 Set 168973_i_at - -
          Murine Genome U74v2 Set 170471_PM_i_at - -
          Murine Genome U74v2 Set 168973_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1419540_PM_at - -
          Mouse Genome 430A 2.0 Array 1419540_PM_at - -
          Mouse Genome 430A 2.0 Array 1419540_PM_at - -
          Mouse Genome 430A 2.0 Array 1419540_at - -
          Mouse Genome 430A 2.0 Array 1419540_at - -
          Mouse Genome 430A 2.0 Array 1419540_PM_at - -
          Mouse Genome 430A 2.0 Array 1419540_at - -
          Mouse Genome 430A 2.0 Array 1419540_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 896952 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550193 - -
          Illumina mouse-6 v1.0 expression beadchip 6550193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215974 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215974 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237603 - -
          mouse-6 v1.1 (Illumina) scl0083457.1_308-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0083457.1_308-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2780736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215974 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2780736 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline