Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00547515_s1
          See other ZSCAN5B GE Assays ›
          Gene Symbol:
          Zscan5b
          Gene Name:
          zinc finger and SCAN domain containing 5B
          Gene Aliases:
          Zfg1, Zfp371
          Chromosome Location:
          Chr.7: 6222278 - 6239417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00547515_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zscan5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133204.2 NP_573467.2 6 1808 141
          GenBank mRNA AF204776.1 - 1 682 141
          AK139659.1 - 6 1809 141

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zscan5b

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          RNA polymerase II transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2419141 AK139659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2419141 BC104342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2419141 AF204776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2419141 NM_133204.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35447 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140276 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2419141 AK139659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2419141 BC104342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2419141 AF204776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2419141 NM_133204.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140276 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06116547 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06116552 - -
          Whole Mouse Genome G4122A A_51_P218095 BC139456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218095 AK139659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218095 BC104342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218095 BC104343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218095 AF204776.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106660 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140276 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06534242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06534247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06534248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06079030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05935741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06652007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06192627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06079033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06643238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06287824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549830 - -
          Affymetrix HT MG-430 PM Array Plate 1449543_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10549827 - -
          Mouse Genome 430 2.0 Array 1449543_PM_at - -
          Mouse Genome 430 2.0 Array 1449543_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170734_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170734_at - -
          Affymetrix Mouse Clariom S Array PSR0700000806.mm.1 NM_133204.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000806.mm.1 NM_133204.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000806.mm.1 TC0700000073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000805.mm.1 NM_133204.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000805.mm.1 NM_133204.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000805.mm.1 TC0700000073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000807.mm.1 NM_133204.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000807.mm.1 NM_133204.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000807.mm.1 TC0700000073.mm.1 MTA TC
          Mouse Expression Set 430 1449543_PM_at - -
          Mouse Expression Set 430 1449543_at - -
          Murine Genome U74v2 Set 134070_PM_at - -
          Murine Genome U74v2 Set 134070_at - -
          Mouse Genome 430A 2.0 Array 1449543_at - -
          Mouse Genome 430A 2.0 Array 1449543_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449543_at - -
          Affymetrix HT Mouse Genome 430A Array 1449543_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 431848 - -
          ABI Mouse Genome Survey Microarray 487678 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5720138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2519822 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2519822 - -
          mouse-6 v1.1 (Illumina) scl0170734.1_214-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0170734.1_214-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2519822 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2519822 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline