| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400031958.mm.1 |
NM_001145958.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031958.mm.1 |
NM_001145958.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031958.mm.1 |
NM_172122.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031958.mm.1 |
NM_172122.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031958.mm.1 |
TC0400003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400031959.mm.1 |
NM_001145958.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031959.mm.1 |
NM_001145958.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031959.mm.1 |
NM_172122.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031959.mm.1 |
NM_172122.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031959.mm.1 |
TC0400003870.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400031957.mm.1 |
NM_001145958.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031957.mm.1 |
NM_001145958.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031957.mm.1 |
NM_172122.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031957.mm.1 |
NM_172122.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400031957.mm.1 |
TC0400003870.mm.1 |
MTA TC |