Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549127_m1
          See other LCORL GE Assays ›
          Gene Symbol:
          Lcorl
          Gene Name:
          ligand dependent nuclear receptor corepressor-like
          Gene Aliases:
          Mlr1
          Chromosome Location:
          Chr.5: 45697184 - 45860115 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549127_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcorl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172153.3 NP_742165.1 1 - 2 208 90
          GenBank mRNA AB076078.1 - 1 - 2 207 90
          AK042567.1 - 1 - 2 209 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002346.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB076078.1,AK042567.1,NM_172153.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcorl

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          DNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_001163073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_001163073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_172153.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_172153.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_178142.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 NM_178142.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021524.mm.1 TC0500002346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500021526.mm.1 NM_172153.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021526.mm.1 NM_172153.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021526.mm.1 TC0500002346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_001163073.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_001163073.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_172153.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_172153.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_178142.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 NM_178142.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021525.mm.1 TC0500002346.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 385238 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline