Search
Search
| Gene Symbol: Lcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB076079.1 | - | 5 - 6 | 423 | 79 | |
| AK041621.1 | - | 5 - 6 | 626 | 79 | ||
| AK077424.1 | - | 5 - 6 | 458 | 79 | ||
| AK129442.1 | - | 3 - 4 | 248 | 79 | ||
| AK155007.1 | - | 5 - 6 | 502 | 79 | ||
| BC090652.1 | - | 2 - 3 | 206 | 30657664 | 79 | |
| BC099938.1 | - | 5 - 6 | 494 | 30085146 | 79 | |
Set Membership: |
|