Search
Search

Pink1| Gene Symbol: Pink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026880.2 | NP_081156.2 | 5 - 6 | 1196 | 79 | |
| GenBank mRNA | AB053476.1 | - | 5 - 6 | 1128 | 79 | |
| AF316872.1 | - | 5 - 6 | 1174 | 79 | ||
| AK050322.1 | - | 5 - 6 | 1176 | 79 | ||
| AK155474.1 | - | 5 - 6 | 1182 | 79 | ||
| AK165480.1 | - | 5 - 6 | 1175 | 79 | ||
| AK220318.1 | - | 5 - 6 | 1150 | 79 | ||
| BC018561.1 | - | 5 - 6 | 1020 | 79 | ||
| BC018594.1 | - | 5 - 6 | 79 | |||
| BC054349.1 | - | 1 - 2 | 79 | |||
| BC067066.1 | - | 5 - 6 | 1164 | 30542123 | 79 | |
| BC069968.1 | - | 6 - 7 | 1054 | 79 | ||
Set Membership: |
|