Search
Search

Pcgf6| Gene Symbol: Pcgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027654.2 | NP_081930.1 | 6 - 7 | 822 | 104 | |
| GenBank mRNA | AB047007.1 | - | 6 - 7 | 872 | 104 | |
| BC016195.1 | - | 6 - 7 | 822 | 4488614 | 104 | |
| BC089460.1 | - | 6 - 7 | 861 | 30632537 | 104 | |
Set Membership: |
|