Search
Search

Polr3f| Gene Symbol: Polr3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029763.3 | NP_084039.2 | 6 - 7 | 696 | 119 | |
| GenBank mRNA | AK029286.1 | - | 6 - 7 | 697 | 119 | |
| AK036166.1 | - | 6 - 7 | 693 | 119 | ||
| AK076538.1 | - | 6 - 7 | 723 | 119 | ||
| AK144679.1 | - | 6 - 7 | 694 | 119 | ||
| BC009159.1 | - | 6 - 7 | 685 | 3602180 | 119 | |
Set Membership: |
|