Search
Search

Slc36a3| Gene Symbol: Slc36a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172258.3 | NP_758462.2 | 7 - 8 | 990 | 79 | |
| GenBank mRNA | AF453745.1 | - | 7 - 8 | 990 | 79 | |
| AK132910.1 | - | 8 - 9 | 1076 | 79 | ||
| AY211261.1 | - | 7 - 8 | 972 | 79 | ||
Set Membership: |
|