Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00552635_m1
          See other ZC3HAV1L GE Assays ›
          Gene Symbol:
          Zc3hav1l
          Gene Name:
          zinc finger CCCH-type, antiviral 1-like
          Gene Aliases:
          B130055L09Rik, E430016P22Rik
          Chromosome Location:
          Chr.6: 38287394 - 38299259 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00552635_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zc3hav1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172467.3 NP_766055.1 3 - 4 796 105
          GenBank mRNA AK045287.1 - 3 - 4 797 105
          AK051064.1 - 3 - 4 798 105
          AK084475.1 - 3 - 4 728 105
          BC138180.1 - 3 - 4 888 8861199 105
          BC138181.1 - 3 - 4 888 8861200 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002098.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zc3hav1l

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376583 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 BC138181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 BC138180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 AK045287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 NM_172467.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 BC138181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 BC138180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 AK045287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P160042 NM_172467.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77264 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119691 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 BC138181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 BC138180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 AK045287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 NM_172467.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 BC138181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 BC138180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 AK045287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P160042 NM_172467.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24376587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119691 - -
          Whole Mouse Genome G4122A A_51_P160042 BC138181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 BC138180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 AK045287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 NM_172467.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 BC138181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 BC138180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 AK045287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P160042 NM_172467.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105509 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05815981 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05224783 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05815980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05224780 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05224782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376581 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376580 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24376582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24376569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05416485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05416492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05416481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05815981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05334183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05833404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05340482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05170096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05170809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05552850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05416478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05416489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24376519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544088 - -
          Affymetrix HT MG-430 PM Array Plate 1456756_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10544084 - -
          Affymetrix Mouse Clariom S Array PSR0600016546.mm.1 TC0600002098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016546.mm.1 NM_172467.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016546.mm.1 NM_172467.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016547.mm.1 TC0600002098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016547.mm.1 NM_172467.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016547.mm.1 NM_172467.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324512 - -
          ABI Mouse Genome Survey Microarray 640667 - -
          ABI Mouse Genome Survey Microarray 775988 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080647 - -
          Illumina mouse-6 v1.0 expression beadchip 5080647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215954 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215954 - -
          mouse-6 v1.1 (Illumina) scl0209032.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0209032.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215954 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215954 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0209032.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline